Learn More
Waters Corp XSelect CSH Phenyl-Hexyl XP HPLC, 2.1 x 50 mm, 2.5 μm
Supplier:  Waters Corp 176002611
Encompass_Preferred
The Waters XSelect CSH Phenyl-Hexyl XP HPLC Column is designed for analytical separations, offering enhanced interaction with planar aromatic compounds. This 2.1 x 50 mm column with 2.5 μm particle size supports robust method development and transferability, providing broad selectivity and high-pressure compatibility.
  • Phenyl phases offer enhanced interaction with planar aromatic compounds via pi-pi interactions.
  • Broad selectivity with superior peak shape for basic compounds and low column bleed.
  • Facilitates robust method development compatible with various chromatographic detection modes and LC system platforms.
  • Scalable to other UPLC and HPLC columns with varying particle sizes and formats.
  • Designed to withstand higher pressures, compatible with HPLC, UHPLC, and UPLC systems.
  • Increases throughput by reducing analysis time through smaller particle technologies.
Catalog No. 50-179-270