Learn More
Waters Corp Waters Column, Phenyl-Hexyl, 130Å, 2.5 μm, 3 mm X 50 mm
Catalog No. 50179534 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns offer an alternative analyte selectivity compared to traditional reversed-phase columns, making them a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions. These columns are packed in ultra-low dispersion hardware.
  • Offers alternative analyte selectivity compared to traditional reversed-phase columns
  • Trifunctionally bonded C6 phenyl-ligand provides a robust, low bleed sorbent
  • Selectively retains polyaromatic compounds through pi-pi interactions
  • Packed in ultra-low dispersion hardware
  • 2.5 μm particles enable exceptional separation performance
  • Provides robustness and throughput for HPLC assays
  • Offers a seamless transition path towards UPLC adoption
  • All column documents are available digitally
Catalog No. 50-179-534 Supplier Waters Corp Supplier No. 186006129
XSelect CSH Phenyl-Hexyl columns offer an alternative analyte selectivity compared to traditional reversed-phase columns, making them a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions. These columns are packed in ultra-low dispersion hardware.
  • Offers alternative analyte selectivity compared to traditional reversed-phase columns
  • Trifunctionally bonded C6 phenyl-ligand provides a robust, low bleed sorbent
  • Selectively retains polyaromatic compounds through pi-pi interactions
  • Packed in ultra-low dispersion hardware
  • 2.5 μm particles enable exceptional separation performance
  • Provides robustness and throughput for HPLC assays
  • Offers a seamless transition path towards UPLC adoption
  • All column documents are available digitally