Learn More
Waters Corp Method Validation Kit, XSelect CSH Phenyl-Hexyl, 130Å, 2.5 μm
Catalog No. 50179609 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect CSH Phenyl-Hexyl Method Validation Kit is designed for rigorous method development and validation. It features a unique phenyl-hexyl phase for enhanced selectivity and includes columns with a pore size of 130Å and particle size of 2.5 μm, making it suitable for a wide range of analytical applications.
  • Unique XSelect CSH Phenyl-Hexyl phase for enhanced selectivity
  • Ideal for method development and validation
  • Pore size of 130Å
  • Particle size of 2.5 μm
  • Column dimensions of 2.1 mm ID x 100 mm length
  • USP L11 classification
Catalog No. 50-179-609 Supplier Waters Corp Supplier No. 186006246
The XSelect CSH Phenyl-Hexyl Method Validation Kit is designed for rigorous method development and validation. It features a unique phenyl-hexyl phase for enhanced selectivity and includes columns with a pore size of 130Å and particle size of 2.5 μm, making it suitable for a wide range of analytical applications.
  • Unique XSelect CSH Phenyl-Hexyl phase for enhanced selectivity
  • Ideal for method development and validation
  • Pore size of 130Å
  • Particle size of 2.5 μm
  • Column dimensions of 2.1 mm ID x 100 mm length
  • USP L11 classification