Learn More
Waters Corp Column, Phenyl-Hexyl, 2.5 μm, 4.6 mm × 75 mm
Catalog No. 50201548 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity, featuring a robust, low bleed C6 phenyl-ligand that selectively retains polyaromatic compounds for method development.
  • Alternative analyte selectivity for method development.
  • Robust, low bleed C6 phenyl-ligand for pi-pi interactions.
  • Ultra-low dispersion hardware; 2.5 μm XP columns for exceptional HPLC separation, robustness, and throughput.
  • Fully compatible with HPLC, UPLC, and UHPLC technologies.
  • Improved performance, backpressure, loading capacity, and particle size scalability over core-shell and fully porous particle columns.
  • Charged Surface Hybrid (CSH) technology enhances LC performance in acidic, low ionic strength.
  • Increases selectivity differences and enhances acidic compound retention (non-end-capped sorbent).
Catalog No. 50-201-548 Supplier Waters Corp Supplier No. 186006134
These XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity, featuring a robust, low bleed C6 phenyl-ligand that selectively retains polyaromatic compounds for method development.
  • Alternative analyte selectivity for method development.
  • Robust, low bleed C6 phenyl-ligand for pi-pi interactions.
  • Ultra-low dispersion hardware; 2.5 μm XP columns for exceptional HPLC separation, robustness, and throughput.
  • Fully compatible with HPLC, UPLC, and UHPLC technologies.
  • Improved performance, backpressure, loading capacity, and particle size scalability over core-shell and fully porous particle columns.
  • Charged Surface Hybrid (CSH) technology enhances LC performance in acidic, low ionic strength.
  • Increases selectivity differences and enhances acidic compound retention (non-end-capped sorbent).