Learn More
Waters Corp Waters OBD Prep Column, C18, 130Å, 5 μm
Supplier:  Waters Corp 186005424
Encompass_Preferred
These CSH C18 columns are general-purpose columns, offering alternative selectivity compared to other C18 sorbents. They utilize Charged Surface Hybrid (CSH) technology to address common column-related issues such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Utilizes Charged Surface Hybrid (CSH) technology.
  • Addresses common column-related issues such as peak shape asymmetry and poor loading for basic compounds.
  • Helps overcome problems like column bleed and slow mobile-phase re-equilibration.
  • Contains hazardous material, requiring special freight handling; additional charges may apply.
Catalog No. 50-791-435
Add to Cart
Request a Quote