Learn More
Waters Corp Waters Corporation MaxPeak Premier OBD Preparative Column, XSelect CSH C18, 50 x 100 mm, 5 μm
Catalog No. 50791467 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
MaxPeak Premier OBD Preparative Columns are designed to minimize component/column surface interactions, thereby reducing non-specific adsorption (NSA) of metal-sensitive compounds and preventing target sample loss. This leads to improved peak shape, sensitivity, and reproducibility. These columns are suitable for reversed-phase purification of drug compounds, peptides, oligonucleotides, or other small molecules.
  • Improved sensitivity and resolution for better compound detection and target purity.
  • Better peak shapes reduce collected fractions and volume.
  • Reduced column conditioning/passivation times save sample, solvent, and time.
  • Improved recovery of target compounds.
  • Minimizes component/column surface interactions and non-specific adsorption (NSA) of metal-sensitive compounds.
Catalog No. 50-791-467 Supplier Waters Corp Supplier No. 186005495
Add to Cart
Add to Cart
Request a Quote
MaxPeak Premier OBD Preparative Columns are designed to minimize component/column surface interactions, thereby reducing non-specific adsorption (NSA) of metal-sensitive compounds and preventing target sample loss. This leads to improved peak shape, sensitivity, and reproducibility. These columns are suitable for reversed-phase purification of drug compounds, peptides, oligonucleotides, or other small molecules.
  • Improved sensitivity and resolution for better compound detection and target purity.
  • Better peak shapes reduce collected fractions and volume.
  • Reduced column conditioning/passivation times save sample, solvent, and time.
  • Improved recovery of target compounds.
  • Minimizes component/column surface interactions and non-specific adsorption (NSA) of metal-sensitive compounds.