Learn More
Waters Corp Waters OBD Prep Column, C18, 5 μm, 130 Å, 19 mm X 250 mm
Catalog No. 50791464 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH C18 columns are general-purpose columns that offer an alternative selectivity compared to other C18 sorbents. They incorporate Charged Surface Hybrid (CSH) technology to address issues such as peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Offers alternative selectivity compared to other C18 sorbents.
  • Utilizes charged surface hybrid (CSH) technology.
  • Helps resolve peak shape asymmetry.
  • Improves loading for basic compounds.
  • Reduces column bleed.
  • Ensures faster mobile-phase re-equilibration.
  • Features spherical particle shape.
  • Provides low silanol activity.
  • Endcapped for enhanced performance.
  • Operates effectively within a pH range of 1 to 11.
  • Withstands maximum pressure up to 6000 psi (415 bar).
  • Maximum temperature of 80 °C (low pH) and 45 °C (high pH).
Catalog No. 50-791-464 Supplier Waters Corp Supplier No. 186005492
Add to Cart
Add to Cart
Request a Quote
XSelect CSH C18 columns are general-purpose columns that offer an alternative selectivity compared to other C18 sorbents. They incorporate Charged Surface Hybrid (CSH) technology to address issues such as peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Offers alternative selectivity compared to other C18 sorbents.
  • Utilizes charged surface hybrid (CSH) technology.
  • Helps resolve peak shape asymmetry.
  • Improves loading for basic compounds.
  • Reduces column bleed.
  • Ensures faster mobile-phase re-equilibration.
  • Features spherical particle shape.
  • Provides low silanol activity.
  • Endcapped for enhanced performance.
  • Operates effectively within a pH range of 1 to 11.
  • Withstands maximum pressure up to 6000 psi (415 bar).
  • Maximum temperature of 80 °C (low pH) and 45 °C (high pH).