Learn More
Waters Corp XSelect Prep Guard Cartridge, 130Å, 5 μm, 19 mm × 10 mm
Catalog No. 50791442 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect CSH Fluoro-Phenyl Prep Guard Cartridge maximizes selectivity differences for method development, especially with low pH mobile phases. Its non-encapped sorbent enhances retention of acidic compounds and protects prep columns from impurities.
  • Maximizes selectivity differences for method development, particularly with low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
  • High-performance guardrail shields prep columns from impurities and contaminants.
  • Preserves lab equipment and samples.
  • Compatible with Acquity UPLC Systems.
  • Advanced 130Å, 5μm sorbent bed for efficient particulate capture.
  • Bonded hybrid silica substrate ensures durability and high-performance.
  • Extends the lifetime of prep columns.
Catalog No. 50-791-442 Supplier Waters Corp Supplier No. 186005431
The XSelect CSH Fluoro-Phenyl Prep Guard Cartridge maximizes selectivity differences for method development, especially with low pH mobile phases. Its non-encapped sorbent enhances retention of acidic compounds and protects prep columns from impurities.
  • Maximizes selectivity differences for method development, particularly with low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
  • High-performance guardrail shields prep columns from impurities and contaminants.
  • Preserves lab equipment and samples.
  • Compatible with Acquity UPLC Systems.
  • Advanced 130Å, 5μm sorbent bed for efficient particulate capture.
  • Bonded hybrid silica substrate ensures durability and high-performance.
  • Extends the lifetime of prep columns.