Learn More
Waters Corp XSelect CSH Fluoro-Phenyl OBD Prep Column, 130Å, 5 µm, 50 mm X 50 mm, 1/pk
Catalog No. 50791473 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.
Catalog No. 50-791-473 Supplier Waters Corp Supplier No. 186005501
Add to Cart
Add to Cart
Request a Quote
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.