Learn More
Waters Corp Column, XSelect CSH Fluoro-Phenyl, 130Å, 5 μm, 50 x 150 mm
Catalog No. 50791475 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect CSH Fluoro-Phenyl OBD Prep Column is a high-performance chromatography column designed for demanding preparative separations. Featuring a 130 Å pore size and 5 μm particle size, this column utilizes a Fluoro-Phenyl phase (PFP) to deliver excellent selectivity and resolution for complex samples. With dimensions of 50 mm ID x 150 mm length, it is suitable for a wide range of analytical applications.
  • High-performance preparative column
  • 130 Å pore size
  • 5 μm particle size
  • Fluoro-Phenyl (PFP) stationary phase
  • Designed for demanding separations
  • Dimensions of 50 mm ID x 150 mm length
  • L43 USP classification
Catalog No. 50-791-475 Supplier Waters Corp Supplier No. 186005503
Add to Cart
Add to Cart
Request a Quote
The XSelect CSH Fluoro-Phenyl OBD Prep Column is a high-performance chromatography column designed for demanding preparative separations. Featuring a 130 Å pore size and 5 μm particle size, this column utilizes a Fluoro-Phenyl phase (PFP) to deliver excellent selectivity and resolution for complex samples. With dimensions of 50 mm ID x 150 mm length, it is suitable for a wide range of analytical applications.
  • High-performance preparative column
  • 130 Å pore size
  • 5 μm particle size
  • Fluoro-Phenyl (PFP) stationary phase
  • Designed for demanding separations
  • Dimensions of 50 mm ID x 150 mm length
  • L43 USP classification