Learn More
Waters Corp XSelect CSH Fluoro-Phenyl OBD Prep Column, 130Å, 5 µm, 19 mm X 100 mm, 1/pk
Catalog No. 50791445 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.
Catalog No. 50-791-445 Supplier Waters Corp Supplier No. 186005434
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.