Learn More
Waters Corp XSelect Preparative Column, CSH Phenyl-Hexyl, 130 Å, 5 μm, 50 x 100 mm
Catalog No. 50791481 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XSelect Preparative Column with CSH Phenyl-Hexyl chemistry provides robust separation capabilities for high-performance liquid chromatography.
  • Offers scalability and versatility for method development, isolation, and purification.
  • Provides rugged performance and exceptional column lifetimes through hybrid materials and OBD Packing Technology.
  • Scalable design reduces solvent consumption and fraction volumes.
  • Revolutionizes high-mass-load preparative separations.
  • Maintains chromatographic peak shape and mass load with formic acid mobile phases.
  • Features CSH Phenyl-Hexyl chemistry for alternative selectivity to C18 and excellent retention for polyaromatic compounds.
  • Ensures reproducibility at pH extremes, delivering superior peak shape and loading capacity for basic compounds.
Catalog No. 50-791-481 Supplier Waters Corp Supplier No. 186005509
Add to Cart
Add to Cart
Request a Quote
This XSelect Preparative Column with CSH Phenyl-Hexyl chemistry provides robust separation capabilities for high-performance liquid chromatography.
  • Offers scalability and versatility for method development, isolation, and purification.
  • Provides rugged performance and exceptional column lifetimes through hybrid materials and OBD Packing Technology.
  • Scalable design reduces solvent consumption and fraction volumes.
  • Revolutionizes high-mass-load preparative separations.
  • Maintains chromatographic peak shape and mass load with formic acid mobile phases.
  • Features CSH Phenyl-Hexyl chemistry for alternative selectivity to C18 and excellent retention for polyaromatic compounds.
  • Ensures reproducibility at pH extremes, delivering superior peak shape and loading capacity for basic compounds.