Learn More
Waters Corp XSelect Prep Column, CSH Phenyl-Hexyl, 130Å, 5 μm, 30 x 75 mm
Catalog No. 50791460 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns offer alternative analyte selectivity compared to traditional reversed-phase columns, making them a valuable tool for method development. The column features a trifunctionally bonded C6 phenyl-ligand, which provides a robust, low bleed sorbent. This sorbent is designed to selectively retain polyaromatic compounds through π–π interactions.
  • Provides alternative analyte selectivity compared to traditional reversed-phase columns.
  • Valuable tool for method development.
  • Trifunctionally bonded C6 phenyl-ligand for robust and low bleed sorbent.
  • Selectively retains polyaromatic compounds through π–π interactions.
Catalog No. 50-791-460 Supplier Waters Corp Supplier No. 186005450
Add to Cart
Add to Cart
Request a Quote
XSelect CSH Phenyl-Hexyl columns offer alternative analyte selectivity compared to traditional reversed-phase columns, making them a valuable tool for method development. The column features a trifunctionally bonded C6 phenyl-ligand, which provides a robust, low bleed sorbent. This sorbent is designed to selectively retain polyaromatic compounds through π–π interactions.
  • Provides alternative analyte selectivity compared to traditional reversed-phase columns.
  • Valuable tool for method development.
  • Trifunctionally bonded C6 phenyl-ligand for robust and low bleed sorbent.
  • Selectively retains polyaromatic compounds through π–π interactions.