Learn More
Waters Corp XSelect CSH Fluoro-Phenyl Column, 130Å, 3.5 µm, 2.1 mm X 75 mm, 1/pk
Supplier:  Waters Corp 186005646
Encompass_Preferred
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.
Catalog No. 50-466-176