Learn More
Waters Corp Waters Method Validation Kit, XSelect HSS C18, 2.5 μm
Supplier: Waters Corp 186006794
This method validation kit includes three HSS C18 columns, each packed with different sorbent batches. Designed for high-performance HPLC and UPLC assays, these 2.5 μm XP columns offer exceptional retention, peak shape, and robust separation in ultra-low dispersion hardware.
- Kit consists of three HSS C18 columns packed with three different batches of sorbent.
- HSS C18 chemistry provides exceptional retention and peak shape.
- 2.5 μm eXtended Performance (XP) columns ensure exceptional separation, robustness, and throughput for HPLC assays.
- Designed for seamless transition to UPLC adoption.
- Chemistry: C18
- Particle size: 2.5 μm
- Pore size: 100 Å
- Inner diameter: 2.1 mm
- Length: 150 mm
- Maximum pressure: 18000 psi (1240 Bar)
- Units per package: 3 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.