Learn More
Waters Corp Waters Corporation Method Validation Kit, C18, 2.5 μm, 3 mm, 150 mm
Catalog No. 50466487 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three columns packed with three different batches of sorbent. It features a general purpose, high performance C18 chemistry, offering exceptional retention and peak shape for various compounds. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness, and throughput for HPLC assays, while facilitating a seamless transition path toward UPLC adoption.
  • Consists of three columns packed with three different batches of sorbent.
  • General purpose, high performance C18 chemistry.
  • Exceptional retention and peak shape for all compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] columns.
  • Enables exceptional separation performance, robustness and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-466-487 Supplier Waters Corp Supplier No. 186006795
This method validation kit consists of three columns packed with three different batches of sorbent. It features a general purpose, high performance C18 chemistry, offering exceptional retention and peak shape for various compounds. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness, and throughput for HPLC assays, while facilitating a seamless transition path toward UPLC adoption.
  • Consists of three columns packed with three different batches of sorbent.
  • General purpose, high performance C18 chemistry.
  • Exceptional retention and peak shape for all compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] columns.
  • Enables exceptional separation performance, robustness and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.