Learn More
Waters Corp XSelect HSS C18 Method Validation Kit, 100Å, 2.5 µm, 4.6 mm X 150 mm, 3/pk
Catalog No. 50466488 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three XSelect HSS C18 columns packed with three different batches of sorbent. XSelect HSS C18 is a general purpose, high performance C18 chemistry with exceptional retention and peak shape for all compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-488 Supplier Waters Corp Supplier No. 186006796
This method validation kit consists of three XSelect HSS C18 columns packed with three different batches of sorbent. XSelect HSS C18 is a general purpose, high performance C18 chemistry with exceptional retention and peak shape for all compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.