Learn More
Waters Corp XSelect Column, C18, 3.5 μm, 100 Å, 1 mm x 50 mm
Catalog No. 50179652 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column utilizes HSS C18 packing, which is a stable bonded phase designed to avoid peak shape issues common in older stationary phases that rely on steric protection or lack of endcapping for low pH stability. It is fully scalable and transferable to UPLC separations.
  • Most stable commercially available bonded phase.
  • Does not suffer from peak shape issues affecting older stationary phases relying on steric protection and lack of endcapping for low pH stability.
  • Fully scalable.
  • Completely transferable to UPLC separations.
Catalog No. 50-179-652 Supplier Waters Corp Supplier No. 186006376
Add to Cart
Add to Cart
This column utilizes HSS C18 packing, which is a stable bonded phase designed to avoid peak shape issues common in older stationary phases that rely on steric protection or lack of endcapping for low pH stability. It is fully scalable and transferable to UPLC separations.
  • Most stable commercially available bonded phase.
  • Does not suffer from peak shape issues affecting older stationary phases relying on steric protection and lack of endcapping for low pH stability.
  • Fully scalable.
  • Completely transferable to UPLC separations.