Learn More
Waters Corp XSelect HSS C18 Column, 100Å, 3.5 µm, 2.1 mm X 100 mm, 1/pk
Supplier:  Waters Corp 186006383
Encompass_Preferred
The HSS C18 packing is the most stable commercially available bonded phase and does not suffer from peak shape issues that plague older stationary phases that rely upon steric protection (and lack of endcapping) to produce stability at low pH. The HSS C18 HPLC columns are fully scalable and are completely transferable to UPLC separations.
Catalog No. 50-179-659