Learn More
Waters Corp Waters Column, C18, 3.5 μm, 2.1 mm x 75 mm
Catalog No. 50179658 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HSS C18 packing is a stable, commercially available bonded phase designed to avoid peak shape issues common with older stationary phases. These columns are fully scalable and transferable to UPLC separations.
  • Superior peak shape for basic chemicals
  • Enhanced retention
  • Outstanding column lifetimes at low pH
  • Minimum column bleed
  • Great batch-to-batch consistency
  • Ultra-efficiencies
  • Seamless column format conversion
  • High Strength Silica (HSS) technology provides improved polar component retention and various selectivity
  • HSS particles offer the mechanical strength required for enduring UPLC pressures
  • Improved resolution for extremely complex samples
  • Improved selectivity for acidic, alkaline, and neutral compounds
  • Improved retention for basic polar compounds when using low-pH mobile phases
Catalog No. 50-179-658 Supplier Waters Corp Supplier No. 186006382
The HSS C18 packing is a stable, commercially available bonded phase designed to avoid peak shape issues common with older stationary phases. These columns are fully scalable and transferable to UPLC separations.
  • Superior peak shape for basic chemicals
  • Enhanced retention
  • Outstanding column lifetimes at low pH
  • Minimum column bleed
  • Great batch-to-batch consistency
  • Ultra-efficiencies
  • Seamless column format conversion
  • High Strength Silica (HSS) technology provides improved polar component retention and various selectivity
  • HSS particles offer the mechanical strength required for enduring UPLC pressures
  • Improved resolution for extremely complex samples
  • Improved selectivity for acidic, alkaline, and neutral compounds
  • Improved retention for basic polar compounds when using low-pH mobile phases