Learn More
Waters Corp Waters Corporation Column, C18, 2.5 μm, 4.6 mm × 150 mm, 100 Å
Catalog No. 50466446 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This general purpose C18 column provides exceptional retention and peak shape for HPLC assays and supports UPLC adoption with 2.5 μm particles and ultra-low dispersion hardware.
  • Separates complex basic and non-basic chemicals at low pH.
  • High Strength Silica (HSS) technology enhances polar molecule retention and selectivity.
  • HSS particles withstand UPLC pressures.
  • Offers alternate analyte selectivity compared to other C18 columns.
  • Fully scalable for UPLC separations.
  • Improves resolution for complex samples.
  • Enhanced selectivity and retention for various compounds with low-pH mobile phases.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Each batch tested for repeatable performance.
  • Efficiency-evaluated columns include an eCord and Certificate of Analysis.
  • Operates effectively up to 10000 psi (690 bar).
Catalog No. 50-466-446 Supplier Waters Corp Supplier No. 186006738
Add to Cart
Add to Cart
This general purpose C18 column provides exceptional retention and peak shape for HPLC assays and supports UPLC adoption with 2.5 μm particles and ultra-low dispersion hardware.
  • Separates complex basic and non-basic chemicals at low pH.
  • High Strength Silica (HSS) technology enhances polar molecule retention and selectivity.
  • HSS particles withstand UPLC pressures.
  • Offers alternate analyte selectivity compared to other C18 columns.
  • Fully scalable for UPLC separations.
  • Improves resolution for complex samples.
  • Enhanced selectivity and retention for various compounds with low-pH mobile phases.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Each batch tested for repeatable performance.
  • Efficiency-evaluated columns include an eCord and Certificate of Analysis.
  • Operates effectively up to 10000 psi (690 bar).