Learn More
Waters Corp Waters Column, C18, 5 μm, 2.1 mm, 50 mm
Supplier:  Waters Corp 186006391
Encompass_Preferred
The HSS C18 packing is the most stable commercially available bonded phase, which avoids peak shape issues that affect older stationary phases relying on steric protection and lack of endcapping for low pH stability. These columns are fully scalable and transferable to UPLC separations.
  • Stable HSS C18 packing for improved peak shape.
  • Fully scalable and transferable to UPLC separations.
  • Wide pH range from 1 to 8.
  • Maximum pressure of 6000 psi (415 bar).
  • 5 μm particle size and 100 Å pore size.
Catalog No. 50-179-667