Learn More
Waters Corp Waters Corporation Column, HSS C18, 5 μm, 3 mm X 150 mm
Supplier:  Waters Corp 186006398
Encompass_Preferred
The HSS C18 packing is the most stable commercially available bonded phase and does not suffer from peak shape issues that plague older stationary phases that rely upon steric protection and lack of endcapping to produce stability at low pH. These HPLC columns are fully scalable and are completely transferable to UPLC separations.
  • Most stable commercially available bonded phase.
  • Avoids peak shape issues common with older stationary phases at low pH.
  • Fully scalable.
  • Completely transferable to UPLC separations.
Catalog No. 50-179-674