Learn More
Waters Corp XSelect HSS C18 Column, 100 Å, 5 μm, 3 mm x 50 mm
Catalog No. 50179672 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Superior peak shape for bases.
  • Exceptional column lifetimes at low pH.
  • Selectivity comparable to modern, fully endcapped C18 columns.
  • HSS technology enhances chromatographic performance.
  • Withstands UPLC pressures; scalable for UPLC separations.
  • Improved polar compound separation and different selectivity.
  • Recommended pH range of 1 to 8.
  • Operates safely up to 6000 psi (415 bar).
Catalog No. 50-179-672 Supplier Waters Corp Supplier No. 186006396
  • Superior peak shape for bases.
  • Exceptional column lifetimes at low pH.
  • Selectivity comparable to modern, fully endcapped C18 columns.
  • HSS technology enhances chromatographic performance.
  • Withstands UPLC pressures; scalable for UPLC separations.
  • Improved polar compound separation and different selectivity.
  • Recommended pH range of 1 to 8.
  • Operates safely up to 6000 psi (415 bar).