Learn More
Waters Corp Waters XSelect HSS C18 Column, 5 μm, 4.6 mm x 30 mm, 100 Å
Catalog No. 50179677 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HSS C18 packing is the most stable commercially available bonded phase and does not suffer from peak shape issues that plague older stationary phases that rely upon steric protection (and lack of endcapping) to produce stability at low pH. The HSS C18 HPLC columns are fully scalable and are completely transferable to UPLC separations.
  • Most stable commercially available bonded phase.
  • Avoids peak shape issues common in older stationary phases relying on steric protection and lack of endcapping for stability at low pH.
  • Fully scalable.
  • Completely transferable to UPLC separations.
Catalog No. 50-179-677 Supplier Waters Corp Supplier No. 186006401
The HSS C18 packing is the most stable commercially available bonded phase and does not suffer from peak shape issues that plague older stationary phases that rely upon steric protection (and lack of endcapping) to produce stability at low pH. The HSS C18 HPLC columns are fully scalable and are completely transferable to UPLC separations.
  • Most stable commercially available bonded phase.
  • Avoids peak shape issues common in older stationary phases relying on steric protection and lack of endcapping for stability at low pH.
  • Fully scalable.
  • Completely transferable to UPLC separations.