Learn More
Waters Corp XSelect HSS C18 Column, 5 μm, 4.6 mm × 75 mm, 100 Å
Catalog No. 50179678 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XSelect HSS C18 Column features a 5 μm particle size and dimensions of 4.6 mm × 75 mm. Constructed from silica, it has a pore size of 100 Å.
  • HSS C18 packing offers a stable bonded phase, avoiding peak shape issues.
  • These HPLC columns are fully scalable and transferable to UPLC separations.
  • Features C18 chemistry and reversed-phase separation mode.
  • Operates within a pH range of 1 to 8.
  • Withstands a maximum pressure of 6000 psi (415 bar).
  • Endcapped with low silanol activity.
Catalog No. 50-179-678 Supplier Waters Corp Supplier No. 186006402
This XSelect HSS C18 Column features a 5 μm particle size and dimensions of 4.6 mm × 75 mm. Constructed from silica, it has a pore size of 100 Å.
  • HSS C18 packing offers a stable bonded phase, avoiding peak shape issues.
  • These HPLC columns are fully scalable and transferable to UPLC separations.
  • Features C18 chemistry and reversed-phase separation mode.
  • Operates within a pH range of 1 to 8.
  • Withstands a maximum pressure of 6000 psi (415 bar).
  • Endcapped with low silanol activity.