Learn More
Waters Corp Waters Corporation Method Validation Kit, XSelect HSS C18, 5 μm, 100 Å
Supplier:  Waters Corp 186006414
Encompass_Preferred
The HSS C18 packing is the most stable commercially available bonded phase. It does not suffer from peak shape issues that affect older stationary phases relying on steric protection and lack of endcapping for low pH stability. The HSS C18 HPLC columns are fully scalable and completely transferable to UPLC separations.
  • Most stable commercially available bonded phase.
  • Avoids peak shape issues common with older stationary phases.
  • Fully scalable to UPLC separations.
  • Completely transferable to UPLC separations.
Catalog No. 50-201-619