Learn More
Waters Corp Method Validation Kit, Trifunctional C18, 2.5 μm, 2.1 x 150 mm
Catalog No. 50466492 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XSelect HSS C18 SB XP Method Validation Kit (MVK) includes a 3-pack of columns, each featuring a Trifunctional C18 stationary phase, a 2.5 μm particle size, and dimensions of 2.1 x 150 mm. This kit is designed for method validation applications.
  • XSelect HSS C18 SB XP Method Validation Kit.
  • Includes 3-pack of columns.
  • Features Trifunctional C18 stationary phase.
  • Each column has 2.5 μm particle size.
  • Column dimensions are 2.1 x 150 mm.
Catalog No. 50-466-492 Supplier Waters Corp Supplier No. 186006800
This XSelect HSS C18 SB XP Method Validation Kit (MVK) includes a 3-pack of columns, each featuring a Trifunctional C18 stationary phase, a 2.5 μm particle size, and dimensions of 2.1 x 150 mm. This kit is designed for method validation applications.
  • XSelect HSS C18 SB XP Method Validation Kit.
  • Includes 3-pack of columns.
  • Features Trifunctional C18 stationary phase.
  • Each column has 2.5 μm particle size.
  • Column dimensions are 2.1 x 150 mm.