Learn More
Waters Corp Waters Column, HSS C18, 3.5 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186006425
Encompass_Preferred
The XSelect HSS C18 column provides exceptional selectivity for chromatographic separations. Utilizing HSS (High Strength Silica) technology, it offers advantages such as increased polar compound retention and diverse selectivity. Designed to withstand UPLC pressures, these columns are seamlessly scalable across various particle sizes and column formats.
  • Increased polar compound retention compared to standard C18 phases.
  • Designed to withstand UPLC pressures.
  • Seamlessly scalable and transferable across different particle sizes and column formats.
  • Provides diverse selectivity for acidic, alkaline, and neutral compounds.
Catalog No. 50-179-690