Learn More
Waters Corp Waters HSS C18 SB Column, 3.5 μm, 2.1 × 75 mm
Catalog No. 50179688 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS C18 SB Column is designed to provide enhanced retention for basic polar compounds when using low-pH mobile phases. It leverages High Strength Silica (HSS) technology for increased polar compound retention and distinct selectivity, while offering the mechanical strength needed to tolerate UPLC pressures.
  • Enhanced retention for basic polar compounds in low-pH mobile phases.
  • High Strength Silica (HSS) technology for distinct selectivity and increased polar compound retention.
  • Mechanical strength to tolerate UPLC pressures.
  • Seamlessly scalable 3.5 μm particles ensure reproducibility with 1.8 μm ACQUITY UPLC HSS.
  • Diverse selectivities, operational stability, and full scalability across LC platforms.
  • Withstands high pressures: up to 18,000 psi (2.1 × 3.0 mm ID) and 9000 psi (4.6 mm ID).
Catalog No. 50-179-688 Supplier Waters Corp Supplier No. 186006423
The XSelect HSS C18 SB Column is designed to provide enhanced retention for basic polar compounds when using low-pH mobile phases. It leverages High Strength Silica (HSS) technology for increased polar compound retention and distinct selectivity, while offering the mechanical strength needed to tolerate UPLC pressures.
  • Enhanced retention for basic polar compounds in low-pH mobile phases.
  • High Strength Silica (HSS) technology for distinct selectivity and increased polar compound retention.
  • Mechanical strength to tolerate UPLC pressures.
  • Seamlessly scalable 3.5 μm particles ensure reproducibility with 1.8 μm ACQUITY UPLC HSS.
  • Diverse selectivities, operational stability, and full scalability across LC platforms.
  • Withstands high pressures: up to 18,000 psi (2.1 × 3.0 mm ID) and 9000 psi (4.6 mm ID).