Learn More
Waters Corp Waters Column, C18, 2.5 μm, 4.6 mm × 150 mm
Supplier: Waters Corp 186006744
This XSelect HSS C18 SB column is designed for low pH separations of complex mixtures (basic and non-basic compounds), offering alternative selectivity to traditional, fully endcapped C18 stationary phases. Its 2.5 μm eXtended Performance (XP) packing in ultra-low dispersion hardware provides excellent separation and robustness for HPLC assays, enabling a clear transition to UPLC.
- C18 chemistry, low coverage, unendcapped.
- Reversed phase separation mode.
- Silica particle substrate.
- pH range of 2 to 8.
- Temperature limit of 45°C.
- Maximum pressure of 10000 psi (690 Bar).
- Particle size of 2.5 μm.
- Pore size of 100 Å.
- Surface area of 230.
- Inner diameter of 4.6 mm.
- Length of 150 mm.
- Carbon load of 8.5%.
- System compatibility with UPLC and UHPLC.
- High silanol activity.
- USP classification L1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.