Learn More
Waters Corp Column, HSS C18 SB, 100Å, 5 μm, 3 mm X 250 mm
Catalog No. 50179705 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HSS C18 SB columns use High Strength Silica (HSS) Technology for excellent peak shape, high efficiency, and robust reversed-phase retention of polar analytes. They offer specific selectivity for bases in challenging liquid chromatography separations. Manufactured in a cGMP, ISO 9001 certified plant, each batch undergoes rigorous chromatographic testing. Features include 100 Å pore diameter, 230 m²/g surface area, pH stability of 2–8, and 8% carbon. Available in 2.5 μm, 3.5 μm, and 5 μm particle sizes.
  • High Strength Silica (HSS) technology for excellent peak shape and efficiency.
  • Specific selectivity for bases in LC separations.
  • pH stability range of 2–8.
  • Particle sizes: 2.5 μm, 3.5 μm, 5 μm; Pore diameter: 100 Å.
  • Manufactured in cGMP, ISO 9001 certified plant.
Catalog No. 50-179-705 Supplier Waters Corp Supplier No. 186006440
These HSS C18 SB columns use High Strength Silica (HSS) Technology for excellent peak shape, high efficiency, and robust reversed-phase retention of polar analytes. They offer specific selectivity for bases in challenging liquid chromatography separations. Manufactured in a cGMP, ISO 9001 certified plant, each batch undergoes rigorous chromatographic testing. Features include 100 Å pore diameter, 230 m²/g surface area, pH stability of 2–8, and 8% carbon. Available in 2.5 μm, 3.5 μm, and 5 μm particle sizes.
  • High Strength Silica (HSS) technology for excellent peak shape and efficiency.
  • Specific selectivity for bases in LC separations.
  • pH stability range of 2–8.
  • Particle sizes: 2.5 μm, 3.5 μm, 5 μm; Pore diameter: 100 Å.
  • Manufactured in cGMP, ISO 9001 certified plant.