Learn More
Waters Corp Waters Column, HSS C18 SB, 100 Å, 5 μm, 3.0 mm × 50 mm
Supplier:  Waters Corp 186006437
Encompass_Preferred
XSelect HPLC Columns are designed for exceptional selectivity, utilizing High Strength Silica (HSS) Technology. They provide high analyte loading, operational stability, and full scalability for UPLC®, HPLC, and preparative separation platforms.
  • High Strength Silica (HSS) Technology offers increased polar compound retention and different selectivity due to the higher silanophilicity of its 100% silica particle.
  • Designed to possess the mechanical strength required to tolerate UPLC pressures.
  • Emphasizes reproducibility and transferability, with 2.5-, 3.5-, and 5-μm HSS HPLC Columns seamlessly scalable with 1.8 μm ACQUITY UPLC HSS particles.
  • Specifically engineered for enhanced retention of basic polar compounds when using low-pH mobile phases.
Catalog No. 50-179-702