Learn More
Waters Corp XSelect HSS C18 SB Column, 100Å, 5 μm, 4.6 mm × 100 mm
Supplier:  Waters Corp 186006444
Encompass_Preferred
The Waters XSelect HSS C18 SB Column is engineered for superior selectivity in base separations, utilizing an un-end-capped, low-coverage C18 bonded phase. It is specifically designed to facilitate efficient low pH separations of complex mixtures containing both basic and non-basic compounds, leveraging High Strength Silica (HSS) technology.
  • Optimized for selectivity of basic compounds.
  • Features an un-end-capped, low-coverage C18 bonded phase.
  • Ideal for low pH separations with complex mixtures.
  • Constructed with High Strength Silica (HSS) technology.
Catalog No. 50-179-709