Learn More
Waters Corp XSelect HSS C18 SB Column, 100 Å, 3.5 μm, 3 mm X 150 mm
Catalog No. 502392509 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HSS C18 SB (Selectivity for Bases) is an unendcapped, low-coverage C18 bonded-phase column, providing increased silanophilic interactions and alternate selectivity for basic compounds compared to fully-endcapped C18 columns. It features an electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history. While compatible with any HPLC system, its advantages are fully realized with the Alliance iS System running Empower software.
  • Unendcapped, low-coverage C18 bonded-phase for increased silanophilic interactions.
  • Provides alternate selectivity for basic compounds.
  • Equipped with an electronic eConnect column tag for unique column-specific information.
  • NFC-enabled for automatic identification, tracking, and digital tracing of column use history.
  • Compatible with any HPLC, with enhanced benefits on Alliance iS System running Empower software.
Catalog No. 50-239-2509 Supplier Waters Corp Supplier No. 186004744RF
Add to Cart
Add to Cart
The HSS C18 SB (Selectivity for Bases) is an unendcapped, low-coverage C18 bonded-phase column, providing increased silanophilic interactions and alternate selectivity for basic compounds compared to fully-endcapped C18 columns. It features an electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history. While compatible with any HPLC system, its advantages are fully realized with the Alliance iS System running Empower software.
  • Unendcapped, low-coverage C18 bonded-phase for increased silanophilic interactions.
  • Provides alternate selectivity for basic compounds.
  • Equipped with an electronic eConnect column tag for unique column-specific information.
  • NFC-enabled for automatic identification, tracking, and digital tracing of column use history.
  • Compatible with any HPLC, with enhanced benefits on Alliance iS System running Empower software.