Learn More
Waters Corp XSelect HSS C18 SB Method Validation Kit, 100Å, 5 µm, 4.6 mm X 100 mm, 3/pk
Supplier:  Waters Corp 186006455
Encompass_Preferred
HSS C18 SB columns are intended for low pH separations that contain complex mixtures of basic and non-basic compounds. HSS C18 SB columns give you alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases. HSS C18 SB HPLC columns are fully scalable and transferable to UPLC separations.
Catalog No. 50-201-630