Learn More
Waters Corp Prep Column, HSS C18 SB, 100Å, 5 μm, 10 mm × 150 mm
Catalog No. 502392927 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HSS C18 SB (Selectivity for Bases) is an unendcapped, low-coverage C18 bonded-phase column providing increased silanophilic interactions for alternate selectivity, particularly for basic compounds.
  • Offers alternate selectivity via increased silanophilic interactions, ideal for basic compounds.
  • Ensures fast, efficient lab-scale separations and high throughput.
  • Facilitates direct scaling from UPLC, UHPLC, or HPLC screening to lab-scale purification.
  • Uses robust chromatographic particles for reliable purification.
  • Patented OBD design provides uniform density, prevents voids, and resists mechanical failure.
  • Delivers consistent performance, extending column life and reducing costs.
  • Maximizes productivity and ensures analytical-to-preparative scalability without method redevelopment.
Catalog No. 50-239-2927 Supplier Waters Corp Supplier No. 186008221
The HSS C18 SB (Selectivity for Bases) is an unendcapped, low-coverage C18 bonded-phase column providing increased silanophilic interactions for alternate selectivity, particularly for basic compounds.
  • Offers alternate selectivity via increased silanophilic interactions, ideal for basic compounds.
  • Ensures fast, efficient lab-scale separations and high throughput.
  • Facilitates direct scaling from UPLC, UHPLC, or HPLC screening to lab-scale purification.
  • Uses robust chromatographic particles for reliable purification.
  • Patented OBD design provides uniform density, prevents voids, and resists mechanical failure.
  • Delivers consistent performance, extending column life and reducing costs.
  • Maximizes productivity and ensures analytical-to-preparative scalability without method redevelopment.