Learn More
Waters Corp XSelect HSS C18 SB XP Column, 100 Å, 2.5 μm, 3.0 × 50 mm
Supplier:  Waters Corp 176002639
Encompass_Preferred
The XSelect HSS C18 SB XP Column is designed for exceptional selectivity and performance in chromatographic separations, utilizing High Strength Silica (HSS) Technology.
  • Enhanced retention for basic polar compounds in low-pH mobile phases.
  • High strength silica (HSS) technology ensures mechanical strength and distinct selectivity.
  • Broad selectivity for polar compounds compared to other C18 phases.
  • XP (eXtended performance) designation for compatibility with HPLC, UHPLC, and UPLC platforms.
  • Robust design to withstand high pressures.
  • Operational stability for reliable results.
Catalog No. 50-179-298