Learn More
Waters Corp XSelect HSS C18 SB XP Column, 100Å, 2.5 µm, 3 mm X 30 mm, 3/pk
Catalog No. 50179297 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS C18 SB column is a low coverage, unendcapped C18 phase used for low pH separations that contain complex mixtures of basic and non-basic compounds. XSelect HSS C18 SB columns give you alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-179-297 Supplier Waters Corp Supplier No. 176002638
The XSelect HSS C18 SB column is a low coverage, unendcapped C18 phase used for low pH separations that contain complex mixtures of basic and non-basic compounds. XSelect HSS C18 SB columns give you alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.