Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, C18 SB, 2.5 μm, 4.6 mm X 100 mm

Catalog No. 50201568 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XSelect HSS C18 SB column is a low coverage, unendcapped C18 phase, designed for low pH separations involving complex mixtures of basic and non-basic compounds. It offers an alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases.

  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
  • C18 chemistry.
  • Particle size: 2.5 μm.
  • Pore size: 100 Å.
  • Inner diameter: 4.6 mm.
  • Length: 100 mm.
  • Maximum pressure: 10000 psi (690 Bar).
  • pH range: 2 to 8.

Catalog No. 50-201-568 Supplier Waters Corp Supplier No. 186006171
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The XSelect HSS C18 SB column is a low coverage, unendcapped C18 phase, designed for low pH separations involving complex mixtures of basic and non-basic compounds. It offers an alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases.

  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
  • C18 chemistry.
  • Particle size: 2.5 μm.
  • Pore size: 100 Å.
  • Inner diameter: 4.6 mm.
  • Length: 100 mm.
  • Maximum pressure: 10000 psi (690 Bar).
  • pH range: 2 to 8.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.