Learn More
Waters Corp VanGuard Cartridge, XSelect HSS C18, 100Å, 3.5 μm
Catalog No. 502391364 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream.
  • Extends analytical column lifetime by removing particulate contamination from the mobile phase.
  • Optimized to protect analytical columns containing 3.5 μm sorbent particles.
  • Features a 100Å, 3.5μm sorbent bed on hybrid silica substrate for impurity retention.
  • Integrates with lab equipment, suitable for diverse applications like protein purification and metabolomics.
  • Improves UPLC system performance with clean mobile phase, enhancing separation efficiency.
  • User-friendly design for simplified installation, compatible with VanGuard Cartridge Holder.
  • Cost-effective solution for column protection, ensuring longevity and consistent performance.
Catalog No. 50-239-1364 Supplier Waters Corp Supplier No. 186007851
VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream.
  • Extends analytical column lifetime by removing particulate contamination from the mobile phase.
  • Optimized to protect analytical columns containing 3.5 μm sorbent particles.
  • Features a 100Å, 3.5μm sorbent bed on hybrid silica substrate for impurity retention.
  • Integrates with lab equipment, suitable for diverse applications like protein purification and metabolomics.
  • Improves UPLC system performance with clean mobile phase, enhancing separation efficiency.
  • User-friendly design for simplified installation, compatible with VanGuard Cartridge Holder.
  • Cost-effective solution for column protection, ensuring longevity and consistent performance.