Learn More
Waters Corp XSelect VanGuard Cartridge, C18, 5 μm, 100 Å, 3.9 mm X 5 mm
Catalog No. 502391355 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect HSS C18 VanGuard Cartridges protect analytical columns by removing particulate contamination. Optimized for 4.6 mm I.D. columns with 5 μm sorbent, they feature HSS (High Strength Silica) for enhanced chromatographic performance, higher polar compound retention, and varied selectivity. These cartridges resist UPLC pressures and are scalable to UPLC separations.
  • C18 chemistry, reversed phase separation
  • Silica spherical particles
  • pH range of 1 to 8
  • Maximum pressure of 18000 psi (1240 Bar)
  • Endcapped with low silanol activity
  • 100 Å pore size
  • 3.9 mm inner diameter, 5 mm length
  • Packaged as 3 units per package
Catalog No. 50-239-1355 Supplier Waters Corp Supplier No. 186007856
XSelect HSS C18 VanGuard Cartridges protect analytical columns by removing particulate contamination. Optimized for 4.6 mm I.D. columns with 5 μm sorbent, they feature HSS (High Strength Silica) for enhanced chromatographic performance, higher polar compound retention, and varied selectivity. These cartridges resist UPLC pressures and are scalable to UPLC separations.
  • C18 chemistry, reversed phase separation
  • Silica spherical particles
  • pH range of 1 to 8
  • Maximum pressure of 18000 psi (1240 Bar)
  • Endcapped with low silanol activity
  • 100 Å pore size
  • 3.9 mm inner diameter, 5 mm length
  • Packaged as 3 units per package