Learn More
Waters Corp Waters XSelect HSS C18 Column, 2.5 μm, 100Å, 3 mm X 50 mm
Catalog No. 50179614 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three batches of HSS C18 columns, each packed with a different batch of sorbent. All column documents are accessible online, as Certificates of Analysis (COAs) and eCords have been removed from select columns to reduce waste and provide easier access.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 1 to 8
  • Maximum pressure of 18000 psi (1240 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 2.5 μm particle size
  • Parker-style endfitting
  • 100 Å pore size
  • Column format
  • 230 surface area
  • Compatible with UPLC and UHPLC systems
  • HSS particle technology
  • L1 USP classification
  • 3 mm inner diameter
  • 50 mm length
  • 15% carbon load
  • Three units per package
Catalog No. 50-179-614 Supplier Waters Corp Supplier No. 186006253
This method validation kit includes three batches of HSS C18 columns, each packed with a different batch of sorbent. All column documents are accessible online, as Certificates of Analysis (COAs) and eCords have been removed from select columns to reduce waste and provide easier access.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 1 to 8
  • Maximum pressure of 18000 psi (1240 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 2.5 μm particle size
  • Parker-style endfitting
  • 100 Å pore size
  • Column format
  • 230 surface area
  • Compatible with UPLC and UHPLC systems
  • HSS particle technology
  • L1 USP classification
  • 3 mm inner diameter
  • 50 mm length
  • 15% carbon load
  • Three units per package