Learn More
Waters Corp XSelect HSS C18 XP Column, 100Å, 2.5 µm, 3 mm X 30 mm, 3/pk
Catalog No. 50179281 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect HSS C18 is a general purpose, high performance C18 chemistry with exceptional retention and peak shape for all compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-179-281 Supplier Waters Corp Supplier No. 176002622
XSelect HSS C18 is a general purpose, high performance C18 chemistry with exceptional retention and peak shape for all compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.