Learn More
Waters Corp XSelect HSS C18 Column, 2.5 µm, 4.6 mm x 75 mm
Catalog No. 50201559 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XSelect HSS C18 column is designed for high-performance liquid chromatography applications. It features a C18 phase and precise dimensions for optimal separation.
  • Phase type C18
  • Dimensions 4.6 mm x 75 mm
  • Particle size 2.5 µm
  • Pore size 100 Å
  • USP L1 application
Catalog No. 50-201-559 Supplier Waters Corp Supplier No. 186006146
This XSelect HSS C18 column is designed for high-performance liquid chromatography applications. It features a C18 phase and precise dimensions for optimal separation.
  • Phase type C18
  • Dimensions 4.6 mm x 75 mm
  • Particle size 2.5 µm
  • Pore size 100 Å
  • USP L1 application