Learn More
Waters Corp Waters XSelect Column, C18, 2.5 μm, 3 mm X 75 mm, 100 Å
Supplier: Waters Corp 176002624
XSelect HSS C18 is a general purpose, high performance C18 chemistry with exceptional retention and peak shape for all compounds. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
- C18 chemistry
- Reversed phase separation mode
- Silica particle substrate
- pH range of 1 to 8 pH
- Maximum pressure up to 18000 psi (1240 Bar)
- Particle size of 2.5 μm
- Pore size of 100 Å
- Compatible with UPLC and UHPLC systems
- HSS particle technology
- Inner diameter of 3 mm
- Length of 75 mm
- Carbon load of 15%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.