Learn More
Waters Corp Waters XSelect HSS C18 XP Column, 2.1 x 30 mm XP, 2.5 μm
Supplier: Waters Corp 176002618
XSelect HPLC XP 2.5 μm Columns offer exceptional separation performance, robustness, and throughput for HPLC assays. These columns are fully compatible with all HPLC, UHPLC, and UPLC Technology platforms, and can improve HPLC productivity by 2 to 4 times with their unmatched selectivity and flexibility.
- Offers exceptional separation performance, robustness, and throughput for HPLC assays.
- Fully compatible with all HPLC, UHPLC, and UPLC technology platforms.
- Improves HPLC productivity by 2 to 4 times with unmatched selectivity and flexibility.
- Directly scalable to other UPLC CSH or UPLC HSS columns and larger XSelect HPLC columns.
- Designed to withstand higher pressures of 9000 psi (for 4.6 mm ID) and 18,000 psi (for 2.1 and 3.0 mm ID).
- Allows users to select column lengths of 30, 50, 75, 100, and 150 mm for optimal resolution and throughput.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.