Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Column, C18, 2.5 μm, 2.1 mm x 75 mm

Catalog No. 50179539 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XSelect HSS C18 is a C18 chemistry that offers exceptional retention and peak shape for all compounds. It is packed in ultra-low dispersion hardware, and the 2.5 μm eXtended Performance (XP) columns provide exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a seamless transition path toward UPLC adoption.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 1 to 8
  • Maximum pressure: 18000 psi (1240 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Particle shape: Spherical
  • Particle size: 2.5 μm
  • Endfitting type: Parker-style
  • Pore size: 100 Å
  • System: UPLC, UHPLC
  • Particle technology: HSS
  • Inner diameter: 2.1 mm
  • Length: 75 mm

Catalog No. 50-179-539 Supplier Waters Corp Supplier No. 186006138
May include imposed supplier surcharges.
Add to Cart
Add to Cart

XSelect HSS C18 is a C18 chemistry that offers exceptional retention and peak shape for all compounds. It is packed in ultra-low dispersion hardware, and the 2.5 μm eXtended Performance (XP) columns provide exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a seamless transition path toward UPLC adoption.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 1 to 8
  • Maximum pressure: 18000 psi (1240 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Particle shape: Spherical
  • Particle size: 2.5 μm
  • Endfitting type: Parker-style
  • Pore size: 100 Å
  • System: UPLC, UHPLC
  • Particle technology: HSS
  • Inner diameter: 2.1 mm
  • Length: 75 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.