Learn More
Waters Corp Waters XSelect HSS C18 XP Column, 3.0 x 50 mm, 2.5 μm
Supplier:  Waters Corp 186006141
Encompass_Preferred
XSelect HPLC Columns, such as the HSS C18 XP, provide a broad, selective, and modern column solution. They feature proprietary High Strength Silica (HSS) technology, offering increased retention for polar compounds and distinct selectivity with 100% silica composition. HSS particles enhance selectivity for acidic, alkaline, and neutral compounds, and are built to tolerate UPLC pressures. The eXtended Performance (XP) 2.5 μm Columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, compatible with HPLC, UHPLC, and UPLC technology platforms.
  • Utilizes HSS technology for increased retention and distinct selectivity.
  • Enhances selectivity for acidic, alkaline, and neutral compounds.
  • Delivers exceptional separation performance, robustness, and throughput.
  • Compatible with HPLC, UHPLC, and UPLC platforms, tolerating pressures up to 18,000 psi.
Catalog No. 50-179-542