Learn More
Waters Corp XSelect HSS C18 Method Validation Kit, 100 Å, 2.5 μm, 4.6 mm x 50 mm
Supplier:  Waters Corp 186006255
Encompass_Preferred
XSelect HSS C18 columns incorporate High Strength Silica (HSS) Technology and are manufactured in a cGMP, ISO 9001 certified plant using ultra-pure reagents. Each column is individually tested and includes a Performance Test Chromatogram and Certificate of Analysis, ensuring reproducible performance for analytical separations.
  • Excellent peak shape.
  • High efficiency.
  • Excellent reversed-phase retention for polar analytes.
  • Recommended operating pH limits are 1–8.
  • Recommended operating temperatures between 20–80 °C.
  • Tolerates operating pressures up to 18,000 psi for 2.5 μm particle size columns.
  • Features a pore diameter of 100 Å.
  • Surface area of 230 m2/g.
  • Ligand density of 3.2 μmol/m2.
  • 15% carbon content.
Catalog No. 50-201-594